siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000534.1 | XP_016856023.1 | 14, 15 | 2749 | ||
| XM_017000535.1 | XP_016856024.1 | 14, 15 | 2709 | |||
| XM_017000536.1 | XP_016856025.1 | 13, 14 | 2557 | |||
| GenBank mRNA | AK296788.1 | - | 13, 14 | 1802 | ||
| BC008655.1 | - | 2, 3 | 311 | |||
| BC037166.2 | - | 14, 15 | 2597 | 5090654 | ||
| M59371.1 | - | 14, 15 | 2592 | |||

EPHA2
Genomic Map