siRNA Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001972.3 | NP_001963.1 | 5 | 835 | ||
| XM_011527775.1 | XP_011526077.1 | 6 | 1512 | |||
| XM_011527776.1 | XP_011526078.1 | 6 | 851 | |||
| GenBank mRNA | BC074816.2 | - | 5 | 752 | 30915534 | |
| BC074817.2 | - | 5 | 752 | 30915320 | ||
| D00187.1 | - | 4 | 625 | |||
| J03545.1 | - | 3 | 493 | |||
| M27783.1 | - | 2 | 349 | |||
| M34379.1 | - | 5 | 753 | |||
| X05875.1 | - | 5 | 753 | |||

ELANE
Genomic Map