siRNA Details
| Gene Symbol: ADGRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256253.1 | NP_001243182.1 | 4 | 381 | ||
| NM_001256255.1 | NP_001243184.1 | 4 | 381 | |||
| NM_001974.4 | NP_001965.3 | 4 | 381 | |||
| XM_011527794.1 | XP_011526096.1 | 4 | 381 | |||
| GenBank mRNA | AK290401.1 | - | 4 | 351 | ||
| AK291518.1 | - | 4 | 351 | |||
| AK297011.1 | - | 4 | 354 | |||
| AK313495.1 | - | 4 | 351 | |||
| BC059395.1 | - | 4 | 364 | 30349201 | ||
| DA465986.1 | - | 4 | 381 | |||
| X81479.1 | - | 4 | 359 | |||
More Information
Set Membership: |
|

ADGRE1
Genomic Map