siRNA Details
Gene Symbol: EN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001427.3 | NP_001418.2 | 2 | 967 | ||
GenBank mRNA | BC104970.1 | - | 2 | 967 | 8143973 | |
BC104972.1 | - | 2 | 967 | 8143975 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001427.3 | NP_001418.2 | 2 | 967 | ||
GenBank mRNA | BC104970.1 | - | 2 | 967 | 8143973 | |
BC104972.1 | - | 2 | 967 | 8143975 |