siRNA Details
| Gene Symbol: EPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004447.5 | NP_004438.3 | 13 | 1598 | ||
| XM_011520605.2 | XP_011518907.1 | 14 | 1658 | |||
| GenBank mRNA | AK291777.1 | - | 13 | 1403 | ||
| AK292931.1 | - | 13 | 1255 | |||
| AK301834.1 | - | 6 | 588 | |||
| AK304860.1 | - | 13 | 1598 | |||
| AK316134.1 | - | 6 | 507 | |||
| AK316239.1 | - | 6 | 780 | |||
| BC030010.2 | - | 13 | 1410 | 4820933 | ||
| U12535.1 | - | 13 | 1370 | |||

EPS8
Genomic Map