siRNA Details
| Gene Symbol: ERH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004450.2 | NP_004441.1 | 3 | 213 | ||
| GenBank mRNA | AK130850.1 | - | 3 | 199 | ||
| AK312186.1 | - | 3 | 208 | |||
| BC014301.2 | - | 3 | 196 | 3507241 | ||
| BC071709.1 | - | 3 | 160 | 5503738 | ||
| BT006877.1 | - | 2 | 142 | |||
| CR450298.1 | - | 2 | 142 | |||
| CR542178.1 | - | 2 | 142 | |||
| D85758.1 | - | 3 | 216 | |||
| U66871.1 | - | 3 | 213 | |||

ERH
Genomic Map