siRNA Details
| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256295.1 | NP_001243224.1 | 10 | 1621 | ||
| NM_005239.5 | NP_005230.1 | 9 | 1451 | |||
| XM_005260935.1 | XP_005260992.1 | 9 | 1524 | |||
| XM_017028290.1 | XP_016883779.1 | 9 | 4121 | |||
| GenBank mRNA | AK096841.1 | - | 10 | 1621 | ||
| AK315563.1 | - | 9 | 1354 | |||
| BC017040.1 | - | 9 | 1323 | 3852274 | ||
| BC042954.2 | - | 9 | 1353 | 5276704 | ||
| BT006838.1 | - | 8 | 1161 | |||
| J04102.1 | - | 9 | 1452 | |||
| M11922.1 | - | 2 | 192 | |||

ETS2
Genomic Map