siRNA Details
| Gene Symbol: EVI2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006495.3 | NP_006486.3 | 2 | 913 | ||
| XM_005257946.3 | XP_005258003.1 | 3 | 1023 | |||
| GenBank mRNA | AK297087.1 | - | 3 | 958 | ||
| BC005926.1 | - | 2 | 874 | 4043339 | ||

EVI2B
Genomic Map