siRNA Details
| Gene Symbol: EYA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005244.4 | NP_005235.3 | 10 | 1298 | ||
| NM_172110.3 | NP_742108.2 | 10 | 1298 | |||
| XM_005260327.2 | XP_005260384.1 | 8 | 745 | |||
| GenBank mRNA | AF055015.1 | - | 6 | 612 | ||
| AK293032.1 | - | 10 | 1002 | |||
| BC000289.2 | - | 4 | 450 | |||
| BC008803.2 | - | 10 | 980 | 3944547 | ||
| BC013882.2 | - | 10 | 1298 | |||
| BT006682.1 | - | 9 | 852 | |||
| U71207.1 | - | 9 | 926 | |||
| U81601.1 | - | 1 | 42 | |||

EYA2
Genomic Map