siRNA Details
| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000130.4 | NP_000121.2 | 13 | 2333 | ||
| XM_017000660.1 | XP_016856149.1 | 12 | 2342 | |||
| GenBank mRNA | AJ297255.1 | - | 1 | 213 | ||
| AK226079.1 | - | 13 | 2251 | |||
| AK291613.1 | - | 13 | 2283 | |||
| CA395604.1 | - | 2 | 278 | |||
| M14335.1 | - | 13 | 2264 | |||
| M16967.1 | - | 13 | 2278 | |||
| M94010.1 | - | 1 | 97 | |||

F5
Genomic Map