siRNA Details
| Gene Symbol: FDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004109.4 | NP_004100.1 | 4 | 689 | ||
| GenBank mRNA | AB096243.1 | - | 4 | 532 | ||
| BC010284.1 | - | 4 | 658 | 3048743 | ||
| BC017063.1 | - | 4 | 639 | 3883924 | ||
| BT006681.1 | - | 4 | 505 | |||
| J03548.1 | - | 4 | 637 | |||
| M18003.1 | - | 4 | 532 | |||
| M34788.1 | - | 4 | 523 | |||

FDX1
Genomic Map