siRNA Details
| Gene Symbol: MLANA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005511.1 | NP_005502.1 | 3 | 200 | ||
| GenBank mRNA | AK312149.1 | - | 3 | 216 | ||
| BC014423.1 | - | 3 | 225 | 4639927 | ||
| CR450299.1 | - | 2 | 147 | |||
| U06452.1 | - | 3 | 200 | |||
| U06654.1 | - | 3 | 307 | |||
| Gene Symbol: KIAA2026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011517759.2 | XP_011516061.1 | 10 | 7863 | ||

MLANA
Genomic Map