siRNA Details
| Gene Symbol: FLNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127487.1 | NP_001120959.1 | 4 | 947 | ||
| NM_001458.4 | NP_001449.3 | 4 | 947 | |||
| GenBank mRNA | AB371585.1 | - | 4 | 967 | ||
| AF089841.1 | - | 4 | 787 | |||
| AF146692.1 | - | 4 | 950 | |||
| AJ012737.1 | - | 4 | 756 | |||

FLNC
Genomic Map