siRNA Details
| Gene Symbol: FRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002031.2 | NP_002022.1 | 7 | 1609 | ||
| XM_005266880.4 | XP_005266937.1 | 8 | 1681 | |||
| XM_005266881.2 | XP_005266938.1 | 8 | 1598 | |||
| XM_005266882.4 | XP_005266939.1 | 9 | 1910 | |||
| XM_011535653.2 | XP_011533955.1 | 9 | 1913 | |||
| XM_011535654.2 | XP_011533956.1 | 8 | 1695 | |||
| XM_011535655.2 | XP_011533957.1 | 8 | 1692 | |||
| XM_011535656.2 | XP_011533958.1 | 7 | 1066 | |||
| XM_017010645.1 | XP_016866134.1 | 8 | 1799 | |||
| GenBank mRNA | AK302264.1 | - | 6 | 784 | ||
| AK313314.1 | - | 7 | 1689 | |||
| BC012916.1 | - | 7 | 1454 | 4515877 | ||
| U00803.1 | - | 7 | 1609 | |||

FRK
Genomic Map