siRNA Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004959.4 | NP_004950.2 | 7 | 1537 | ||
| XM_005251871.3 | XP_005251928.1 | 7 | 1565 | |||
| XM_011518455.2 | XP_011516757.1 | 7 | 1415 | |||
| GenBank mRNA | AB307718.1 | - | 6 | 1350 | ||
| AK090858.1 | - | 2 | 983 | |||
| BC032501.1 | - | 7 | 1537 | 5227690 | ||
| HQ692856.1 | - | 6 | 1350 | |||
| HQ709184.1 | - | 5 | 1230 | |||
| U76388.1 | - | 6 | 1365 | |||

NR5A1
Genomic Map