siRNA Details
| Gene Symbol: GABRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000812.3 | NP_000803.2 | 8 | 1373 | ||
| XM_017007985.1 | XP_016863474.1 | 4 | 511 | |||
| GenBank mRNA | AK225167.1 | - | 8 | 1050 | ||
| AK225686.1 | - | 8 | 1077 | |||
| AK293691.1 | - | 7 | 950 | |||
| AK312720.1 | - | 8 | 1019 | |||
| BC022449.1 | - | 8 | 1077 | 4797401 | ||
| KJ534841.1 | - | 8 | 999 | |||
| KJ535054.1 | - | 7 | 931 | |||
| X14767.1 | - | 8 | 1030 | |||

GABRB1
Genomic Map