siRNA Details
| Gene Symbol: GABRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000812.3 | NP_000803.2 | 5 | 905 | ||
| GenBank mRNA | AK225167.1 | - | 5 | 582 | ||
| AK293691.1 | - | 4 | 482 | |||
| AK312720.1 | - | 5 | 551 | |||
| BC022449.1 | - | 5 | 609 | 4797401 | ||
| KJ534841.1 | - | 5 | 531 | |||
| KJ535054.1 | - | 4 | 463 | |||
| X14767.1 | - | 5 | 562 | |||

GABRB1
Genomic Map