siRNA Details
| Gene Symbol: KAT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021078.2 | NP_066564.2 | 9 | 1439 | ||
| XM_006721817.3 | XP_006721880.1 | 9 | 1468 | |||
| XM_006721818.3 | XP_006721881.1 | 9 | 1351 | |||
| XM_017024448.1 | XP_016879937.1 | 9 | 1602 | |||
| GenBank mRNA | AF029777.1 | - | 9 | 1403 | ||
| BC032743.1 | - | 9 | 1416 | 5575574 | ||
| BC039907.1 | - | 9 | 1431 | 6064838 | ||
| BC105977.1 | - | 9 | 1408 | 6137532 | ||

KAT2A
Genomic Map