siRNA Details
| Gene Symbol: GDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016204.2 | NP_057288.1 | 2 | 568 | ||
| GenBank mRNA | AF188285.1 | - | 2 | 552 | ||
| AK314956.1 | - | 2 | 568 | |||
| BC069643.1 | - | 2 | 471 | 7262433 | ||
| BC074921.2 | - | 2 | 420 | 30915569 | ||

GDF2
Genomic Map