siRNA Details
| Gene Symbol: GIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005142.2 | NP_005133.2 | 5 | 582 | ||
| XM_011544939.2 | XP_011543241.1 | 5 | 610 | |||
| GenBank mRNA | AK301295.1 | - | 5 | 694 | ||
| AK314275.1 | - | 5 | 601 | |||
| BC037958.1 | - | 5 | 582 | 5759934 | ||
| X76562.1 | - | 5 | 581 | |||

GIF
Genomic Map