siRNA Details
| Gene Symbol: GLI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000168.5 | NP_000159.3 | 15 | 3855 | ||
| XM_011515274.2 | XP_011513576.1 | 14 | 6479 | |||
| XM_017011997.1 | XP_016867486.1 | 14 | 6512 | |||
| GenBank mRNA | AK299299.1 | - | 14 | 3741 | ||
| BC113616.1 | - | 14 | 3812 | 8322668 | ||
| BC117168.1 | - | 14 | 3812 | 40125719 | ||
| M57609.1 | - | 14 | 3818 | |||

GLI3
Genomic Map