siRNA Details
Gene Symbol: GNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012202.4 | NP_036334.1 | 3 | 452 | ||
XM_006718500.2 | XP_006718563.1 | 4 | 439 | |||
GenBank mRNA | AF075042.1 | - | 2 | 115 | ||
AF087900.1 | - | 3 | 389 | |||
AF092129.1 | - | 3 | 389 | |||
AF493871.1 | - | 2 | 129 | |||
AK311933.1 | - | 3 | 352 | |||
BC015563.2 | - | 3 | 333 | 4124714 | ||
CK004020.1 | - | 3 | 406 | |||
DA123305.1 | - | 3 | 452 |
