siRNA Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002077.3 | NP_002068.1 | 12 | 1344 | ||
| XM_005251929.3 | XP_005251986.1 | 12 | 1276 | |||
| XM_006717062.3 | XP_006717125.1 | 12 | 1340 | |||
| XM_006717063.3 | XP_006717126.1 | 12 | 1311 | |||
| GenBank mRNA | AB208858.1 | - | 7 | 701 | ||
| AK302445.1 | - | 12 | 1153 | |||
| BC032853.1 | - | 12 | 1314 | 4828793 | ||
| U51587.1 | - | 12 | 1336 | |||

GOLGA1
Genomic Map