siRNA Details
Gene Symbol: GPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005281.3 | NP_005272.1 | 2 | 149 | ||
GenBank mRNA | AK291185.1 | - | 2 | 136 | ||
AK314868.1 | - | 2 | 128 | |||
BC032702.1 | - | 2 | 145 | 5247608 | ||
BQ882813.1 | - | 2 | 149 | |||
HQ709189.1 | - | 1 | 50 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
