siRNA Details
| Gene Symbol: GPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005282.2 | NP_005273.1 | 2 | 1176 | ||
| XM_017026607.1 | XP_016882096.1 | 2 | 1764 | |||
| XM_017026608.1 | XP_016882097.1 | 2 | 1085 | |||
| GenBank mRNA | AK290698.1 | - | 2 | 1129 | ||
| BC038450.1 | - | 2 | 1164 | |||
| BC067535.1 | - | 1 | 535 | 6971751 | ||
| BC067536.1 | - | 1 | 535 | 6971752 | ||
| EU432116.1 | - | 1 | 231 | |||

GPR4
Genomic Map