siRNA Details
| Gene Symbol: GPR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161415.1 | NP_001154887.1 | 4 | 806 | ||
| NM_001161416.1 | NP_001154888.1 | 3 | 741 | |||
| NM_001161417.1 | NP_001154889.1 | 2 | 512 | |||
| NM_005291.2 | NP_005282.1 | 3 | 577 | |||
| XM_017003833.1 | XP_016859322.1 | 2 | 933 | |||
| GenBank mRNA | AK126849.1 | - | 1 | 1767 | ||
| AK292725.1 | - | 2 | 382 | |||
| AK313949.1 | - | 2 | 268 | |||
| BC039595.1 | - | 2 | 196 | |||
| DA154324.1 | - | 2 | 512 | |||
| DA314295.1 | - | 3 | 289 | |||
| DA390892.1 | - | 2 | 491 | |||
| JF810891.1 | - | 1 | 111 | |||
| Y12546.1 | - | 3 | 269 | |||
| Z94154.1 | - | 2 | 205 | |||

GPR17
Genomic Map