siRNA Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005295.2 | NP_005286.2 | 3 | 2510 | ||
| XM_011516056.2 | XP_011514358.1 | 2 | 2196 | |||
| GenBank mRNA | AB209270.1 | - | 1 | 1420 | ||
| AK122621.1 | - | 3 | 2507 | |||
| AK307393.1 | - | 3 | 2507 | |||
| BC107128.2 | - | 1 | 1164 | 40024013 | ||
| BX647649.1 | - | 2 | 1374 | |||
| HQ995527.1 | - | 1 | 1164 | |||
More Information
Set Membership: |
|

GPR22
Genomic Map