siRNA Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278000.1 | NP_001264929.1 | 5 | 797 | ||
| NM_005296.2 | NP_005287.1 | 2 | 582 | |||
| XM_005262126.3 | XP_005262183.1 | 5 | 1000 | |||
| XM_006724639.3 | XP_006724702.1 | 4 | 712 | |||
| XM_017029437.1 | XP_016884926.1 | 5 | 2059 | |||
| XM_017029438.1 | XP_016884927.1 | 3 | 735 | |||
| GenBank mRNA | AK314134.1 | - | 2 | 576 | ||
| BC069996.1 | - | 1 | 238 | 6971739 | ||
| BC074722.2 | - | 1 | 202 | 30915172 | ||
| BC095538.1 | - | 1 | 351 | 30706564 | ||
| U90322.1 | - | 1 | 243 | |||

LPAR4
Genomic Map