siRNA Details
| Gene Symbol: GRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000841.3 | NP_000832.1 | 2 | 936 | ||
| NM_001256811.2 | NP_001243740.1 | 2 | 936 | |||
| NM_001282847.1 | NP_001269776.1 | 2 | 936 | |||
| XM_017010790.1 | XP_016866279.1 | 2 | 874 | |||
| XM_017010791.1 | XP_016866280.1 | 2 | 929 | |||
| GenBank mRNA | AK289524.1 | - | 1254 | |||
| AK289534.1 | - | 2 | 879 | |||
| AK308669.1 | - | 2 | 404 | |||
| AK312300.1 | - | 2 | 891 | |||
| BC130526.1 | - | 1 | 594 | 40146557 | ||
| BC130528.1 | - | 1 | 594 | 40146559 | ||
| BC144057.1 | - | 1 | 594 | 9052577 | ||
| EU432124.1 | - | 1 | 435 | |||
| U92457.1 | - | 1 | 460 | |||
| X80818.1 | - | 1 | 605 | |||
More Information
Set Membership: |
|

GRM4
Genomic Map