siRNA Details
| Gene Symbol: GYS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021957.3 | NP_068776.2 | 1 | 349 | ||
| XM_017019245.1 | XP_016874734.1 | 1 | 360 | |||
| GenBank mRNA | AK308373.1 | - | 1 | 321 | ||
| BC126310.1 | - | 1 | 119 | 8992026 | ||
| BC126312.1 | - | 1 | 119 | 8992028 | ||
| D29685.1 | - | 1 | 348 | |||
| DB185936.1 | - | 1 | 360 | |||
| S70004.1 | - | 1 | 122 | |||

GYS2
Genomic Map