siRNA Details
| Gene Symbol: H1F0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005318.3 | NP_005309.1 | 1 | 732 | ||
| GenBank mRNA | AK312583.1 | - | 1 | 598 | ||
| BC000145.1 | - | 1 | 579 | 3049108 | ||
| BC029046.1 | - | 1 | 579 | 5189077 | ||
| CR456502.1 | - | 1 | 410 | |||
| CR542220.1 | - | 1 | 294 | |||

H1F0
Genomic Map