siRNA Details
| Gene Symbol: HIST1H1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005320.2 | NP_005311.1 | 1 | 182 | ||
| GenBank mRNA | AK312845.1 | - | 1 | 181 | ||
| BC104874.1 | - | 1 | 208 | 8143877 | ||
| BC111971.1 | - | 1 | 208 | 8327439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H1D| Gene Symbol: HIST1H1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005320.2 | NP_005311.1 | 1 | 182 | ||
| GenBank mRNA | AK312845.1 | - | 1 | 181 | ||
| BC104874.1 | - | 1 | 208 | 8143877 | ||
| BC111971.1 | - | 1 | 208 | 8327439 | ||