siRNA Details
| Gene Symbol: HIST1H1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005325.3 | NP_005316.1 | 1 | 365 | ||
| GenBank mRNA | BC069492.1 | - | 1 | 336 | 7262187 | |
| BC101593.1 | - | 1 | 364 | 8069099 | ||
| BC112140.1 | - | 1 | 365 | 8327608 | ||

HIST1H1A
Genomic Map