siRNA Details
| Gene Symbol: HLA-DQA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020056.4 | NP_064440.1 | 2 | 274 | ||
| GenBank mRNA | M95727.1 | - | 1 | 114 | ||
| M95728.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HLA-DQA2| Gene Symbol: HLA-DQA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020056.4 | NP_064440.1 | 2 | 274 | ||
| GenBank mRNA | M95727.1 | - | 1 | 114 | ||
| M95728.1 | - | 1 | 114 | |||