siRNA Details
| Gene Symbol: HOXB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002147.3 | NP_002138.1 | 1 | 215 | ||
| GenBank mRNA | AK314964.1 | - | 1 | 272 | ||
| BC117247.1 | - | 1 | 176 | 40125798 | ||
| BE621135.1 | - | 1 | 13 | |||
| BQ073272.1 | - | 149 | ||||
| M92299.1 | - | 1 | 431 | |||

HOXB5
Genomic Map