siRNA Details
Gene Symbol: HOXC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022658.3 | NP_073149.1 | 1 | 432 | ||
GenBank mRNA | AK290959.1 | - | 1 | 423 | ||
BC053898.1 | - | 1 | 432 | 6171090 | ||
BU156590.1 | - | 443 | ||||
X99680.1 | - | 1 | 64 |
