siRNA Details
Gene Symbol: HOXC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000445.1 | - | 2 | 672 | ||
AK315194.1 | - | 2 | 689 | |||
BC032769.1 | - | 2 | 663 | |||
BC053894.1 | - | 2 | 724 | 6144625 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOXC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000445.1 | - | 2 | 672 | ||
AK315194.1 | - | 2 | 689 | |||
BC032769.1 | - | 2 | 663 | |||
BC053894.1 | - | 2 | 724 | 6144625 |