siRNA Details
| Gene Symbol: HOXC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017410.2 | NP_059106.2 | 2 | 1081 | ||
| GenBank mRNA | AF255676.1 | - | 2 | 1081 | ||
| AF263466.2 | - | 2 | 1038 | |||
| AK024027.1 | - | 2 | 563 | |||
| BC002754.1 | - | 2 | 1028 | |||
| BC090850.1 | - | 2 | 1050 | 6171228 | ||
| BT009908.1 | - | 2 | 966 | |||
| BU158600.1 | - | 2 | 370 | |||

HOXC13
Genomic Map