siRNA Details
| Gene Symbol: HOXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014621.2 | NP_055436.2 | 2 | 1002 | ||
| XM_005246514.4 | XP_005246571.1 | 2 | 1896 | |||
| GenBank mRNA | AK311735.1 | - | 2 | 432 | ||
| AK313885.1 | - | 2 | 922 | |||
| BC016763.1 | - | 2 | 820 | 4068219 | ||
| BC074797.2 | - | 2 | 800 | 30915295 | ||
| X04706.1 | - | 2 | 1002 | |||

HOXD4
Genomic Map