siRNA Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000413.2 | NP_000404.2 | 1 | 1048 | ||
| XM_005257292.3 | XP_005257349.1 | 1 | 715 | |||
| XM_006721857.3 | XP_006721920.1 | 1 | 715 | |||
| XM_006721858.3 | XP_006721921.1 | 1 | 715 | |||
| XM_006721859.3 | XP_006721922.1 | 1 | 715 | |||
| XM_011524729.1 | XP_011523031.1 | 1 | 456 | |||
| XM_011524730.1 | XP_011523032.1 | 1 | 456 | |||
| XM_011524731.1 | XP_011523033.1 | 1 | 456 | |||
| XM_011524732.2 | XP_011523034.1 | 1 | 715 | |||
| GenBank mRNA | AK127832.1 | - | 1 | 3800 | ||
| AK300143.1 | - | 1 | 91 | |||
| AK300159.1 | - | 1 | 93 | |||
| AK300452.1 | - | 1 | 91 | |||
| AK316292.1 | - | 1 | 276 | |||
| BC033110.1 | - | 1 | 520 | |||
| BC037415.1 | - | 1 | 1694 | |||
| BC104752.1 | - | 1 | 106 | 8143755 | ||
| BC111935.1 | - | 1 | 106 | 8327403 | ||
| DQ469954.1 | - | 1 | 80 | |||
| M36263.1 | - | 1 | 89 | |||
| X13440.1 | - | 1 | 89 | |||

HSD17B1
Genomic Map