siRNA Details
Gene Symbol: HTR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000865.2 | NP_000856.1 | 2 | 1593 | ||
XM_011535789.2 | XP_011534091.1 | 2 | 2041 | |||
GenBank mRNA | AF498980.1 | - | 1 | 890 | ||
BC069751.1 | - | 2 | 1172 | 7262611 | ||
M91467.1 | - | 3 | 1456 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
