siRNA Details
| Gene Symbol: IFNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005534.3 | NP_005525.2 | 2 | 822 | ||
| XM_005260969.2 | XP_005261026.1 | 3 | 338 | |||
| XM_011529553.1 | XP_011527855.1 | 3 | 264 | |||
| XM_011529554.2 | XP_011527856.1 | 2 | 698 | |||
| GenBank mRNA | AK292246.1 | - | 2 | 270 | ||
| AK314005.1 | - | 2 | 283 | |||
| BC003624.2 | - | 2 | 290 | 2967074 | ||
| CB992729.1 | - | 2 | 332 | |||
| EF173886.1 | - | 1 | 55 | |||
| U05875.1 | - | 2 | 822 | |||
| U05877.1 | - | 2 | 287 | |||

IFNGR2
Genomic Map