siRNA Details
| Gene Symbol: JCHAIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144646.3 | NP_653247.1 | 4 | 560 | ||
| XM_006714211.2 | XP_006714274.1 | 5 | 525 | |||
| XM_006714212.2 | XP_006714275.1 | 5 | 462 | |||
| XM_011531925.1 | XP_011530227.1 | 5 | 462 | |||
| XM_011531926.1 | XP_011530228.1 | 5 | 462 | |||
| GenBank mRNA | AK312014.1 | - | 4 | 450 | ||
| BC038982.1 | - | 4 | 476 | 5755337 | ||
| BX648421.1 | - | 1 | 2020 | |||
| DQ884395.1 | - | 5 | 467 | |||

JCHAIN
Genomic Map