siRNA Details
| Gene Symbol: IHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002181.3 | NP_002172.2 | 1 | 307 | ||
| GenBank mRNA | BC136587.1 | - | 1 | 307 | 9020577 | |
| BC136588.1 | - | 1 | 307 | 9020578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IHH| Gene Symbol: IHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002181.3 | NP_002172.2 | 1 | 307 | ||
| GenBank mRNA | BC136587.1 | - | 1 | 307 | 9020577 | |
| BC136588.1 | - | 1 | 307 | 9020578 | ||