siRNA Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000882.3 | NP_000873.2 | 7 | 949 | ||
| GenBank mRNA | AF101062.1 | - | 7 | 728 | ||
| AF180562.1 | - | 7 | 787 | |||
| BC104982.1 | - | 7 | 881 | 8143985 | ||
| BC104984.1 | - | 7 | 881 | 8143987 | ||
| M65291.1 | - | 7 | 829 | |||

IL12A
Genomic Map