siRNA Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243211.1 | NP_001230140.1 | 2 | 234 | ||
| NM_001562.3 | NP_001553.1 | 2 | 234 | |||
| XM_011542805.1 | XP_011541107.1 | 2 | 242 | |||
| XM_011542806.2 | XP_011541108.1 | 2 | 238 | |||
| GenBank mRNA | AF077611.1 | - | 1 | 14 | ||
| AF380360.1 | - | 2 | 66 | |||
| AK311301.1 | - | 2 | 210 | |||
| AY044641.1 | - | 2 | 111 | |||
| AY266351.1 | - | 2 | 191 | |||
| BC007007.1 | - | 2 | 209 | 3686124 | ||
| BC007461.1 | - | 2 | 234 | 3681009 | ||
| CR541973.1 | - | 1 | 14 | |||
| CR542001.1 | - | 1 | 14 | |||
| D49950.1 | - | 2 | 191 | |||
| U90434.1 | - | 11 | ||||

IL18
Genomic Map