siRNA Details
| Gene Symbol: IMPDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000884.2 | NP_000875.2 | 12 | 1446 | ||
| XM_006713128.3 | XP_006713191.1 | 13 | 1826 | |||
| XM_017006349.1 | XP_016861838.1 | 12 | 1703 | |||
| XM_017006350.1 | XP_016861839.1 | 11 | 1631 | |||
| GenBank mRNA | BC006124.1 | - | 12 | 1394 | 3545693 | |
| BC012840.1 | - | 12 | 1387 | 3447994 | ||
| BC015567.2 | - | 12 | 1385 | 4576285 | ||
| J04208.1 | - | 12 | 1401 | |||

IMPDH2
Genomic Map