siRNA Details
| Gene Symbol: IMPDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000884.2 | NP_000875.2 | 3, 4 | 342 | ||
| XM_006713128.3 | XP_006713191.1 | 3, 4 | 650 | |||
| GenBank mRNA | AK293397.1 | - | 303 | |||
| BC006124.1 | - | 3, 4 | 290 | 3545693 | ||
| BC012840.1 | - | 3, 4 | 283 | 3447994 | ||
| BC015567.2 | - | 3, 4 | 281 | 4576285 | ||
| J04208.1 | - | 3, 4 | 297 | |||

IMPDH2
Genomic Map