siRNA Details
| Gene Symbol: ING2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001564.3 | NP_001555.1 | 1 | 278 | ||
| GenBank mRNA | AB012853.1 | - | 1 | 223 | ||
| AF053537.1 | - | 1 | 199 | |||
| AJ006851.1 | - | 1 | 132 | |||
| AK314000.1 | - | 1 | 279 | |||
| BC030128.2 | - | 1 | 258 | 3941655 | ||

ING2
Genomic Map