siRNA Details
| Gene Symbol: ISL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002202.2 | NP_002193.2 | 5 | 1410 | ||
| XM_011543380.2 | XP_011541682.1 | 4 | 2528 | |||
| GenBank mRNA | AK290742.1 | - | 5 | 1128 | ||
| BC031213.1 | - | 5 | 1411 | 5269369 | ||
| S70721.1 | - | 5 | 867 | |||
| U07559.1 | - | 5 | 1101 | |||

ISL1
Genomic Map